missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(II) sulfide, tech. 90%, Thermo Scientific Chemicals

Product Code. 11323337
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Arsenic(II) sulfide is used as an primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-79-0
Molecular Formula AsH3S
Molecular Weight (g/mol) 110.01
MDL Number MFCD00049841
InChI Key BMOFEAKQXGGUPE-UHFFFAOYSA-N
Synonym arsenic ii sulfide, realgar ass, diarsanedithione
IUPAC Name arsanethiol
SMILES S[AsH2]

Specifications

Boiling Point 565°C
Melting Point ∼307°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Solubility Information Insoluble in water. Soluble in acids and alkalis.
Formula Weight 213.97
Percent Purity 90%
Grade Technical
Odor Odorless
Density 3.5 g/mL
Chemical Name or Material Arsenic(II) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'