Learn More
Arsenic(III) iodide, 98%, Thermo Scientific Chemicals
£118.11 - £405.02
Chemical Identifiers
CAS | 7784-45-4 |
---|---|
Molecular Formula | AsI3 |
Molecular Weight (g/mol) | 455.635 |
MDL Number | MFCD00014170 |
InChI Key | IKIBSPLDJGAHPX-UHFFFAOYSA-N |
Synonym | arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid |
PubChem CID | 24575 |
IUPAC Name | triiodoarsane |
SMILES | [As](I)(I)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15462127
|
Thermo Scientific Alfa Aesar
045067.09 |
10 g |
£118.11
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472127
|
Thermo Scientific Alfa Aesar
045067.18 |
50 g |
£405.02
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arsenic(III) iodide is mainly used to treat rheumatism, arthritis, malaria, trypanosome infections, tuberculosis, and diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-45-4 | |
455.635 | |
IKIBSPLDJGAHPX-UHFFFAOYSA-N | |
24575 | |
[As](I)(I)I |
AsI3 | |
MFCD00014170 | |
arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodidShow More | |
triiodoarsane |
Specifications
424°C | |
7784-45-4 | |
AsI3 | |
UN1557 | |
arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid | |
IKIBSPLDJGAHPX-UHFFFAOYSA-N | |
triiodoarsane | |
24575 | |
98% | |
Moisture sensitive | |
Arsenic (III) iodide |
140°C | |
Pieces | |
MFCD00014170 | |
14,803 | |
Soluble in alcohol,ether,Carbon disulfide,water(6g/100 m). | |
[As](I)(I)I | |
455.635 | |
455.64 | |
Pungent | |
4.39 g/mL |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 232-068-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.