missing translation for 'onlineSavingsMsg'
Learn More
Arsenic(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11353938

Arsenic(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11353938
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11353938

Brand: Thermo Scientific Alfa Aesar 040370.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 465°C (sublimation)
Melting Point 312.3°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in dilute acids and alkalies.Insoluble in organic solvents.Slightly soluble in water.
Formula Weight 197.84
Odor Odorless
Density 3.74
Chemical Name or Material Arsenic(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ17K006 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number040370.18,040370.09,040370.30
Lot NumberY14J040 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number040370.18,040370.09,040370.30
Lot NumberQ18C008 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number040370.18,040370.09,040370.30
Lot NumberM12H027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040370.18,040370.09,040370.30
Lot NumberZ22H012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040370.18,040370.09,040370.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic(III) oxide, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.