missing translation for 'onlineSavingsMsg'
Learn More
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals Product Code.: 11346188

Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals

Product Code. 11346188
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11346188

Brand: Thermo Scientific Alfa Aesar 011173.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 465°C (sublimation)
Melting Point 312.3°C
Physical Form Powder
Quantity 10 g
UN Number UN1561
Merck Index 14,804
Solubility Information Soluble in hot water,dilute acids and alkalies. Insoluble in alcohol,chloroform,and ether,.
Formula Weight 197.84
Percent Purity 99.996%
Grade Puratronic™
Odor Odorless
Density 3.74 g/mL
Assay (metals basis excluding Sb), Sb 20ppm max.
Chemical Name or Material Arsenic(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25L014 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number011173.18,011173.09,011173.04
Lot NumberY11K043 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number011173.18,011173.09,011173.04
Lot NumberU16K030 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number011173.18,011173.09,011173.04
Lot NumberZ29I013 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number011173.18,011173.09,011173.04
Lot NumberN18H047 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011173.18,011173.09,011173.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.