missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
5 g
25 g
Description
Arsenicselenide is used as a chalcogenide glass for infrared optics, as it transmits light with wavelengths between 870 nm and 17.2 μm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 260°C |
Physical Form | Fused Pieces |
Quantity | 5 g |
UN Number | UN3077 |
Merck Index | 14,805 |
Solubility Information | Insoluble in water and soluble in nitric acid. |
Formula Weight | 386.72 |
Percent Purity | 99.999% |
Odor | Odorless |
Density | 4.75 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'