missing translation for 'onlineSavingsMsg'
Learn More
Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15442417

Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15442417
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15442417

Brand: Thermo Scientific Alfa Aesar 045616.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Arsenic(III) sulfide is used in the treatment of acute promyelocytic leukemia. It is an inorganic photoresist used in the fabrication of photonic crystals and 3-D nanostructures. It is also used as an acousto-optic material, pigment, tanning agent and chalcogenide glass for infrared optics. Further, it is used as an analytical test for presence of dissimilatory arsenic-reducing bacteria (DARB).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]

Specifications

Boiling Point 707°C
Melting Point 300°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Merck Index 14,806
Solubility Information Insoluble in water.
Formula Weight 246.03
Percent Purity 99.9%
Odor Like rotten eggs
Density 3.43 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic (III) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN26L009 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog Number045616.06,045616.14
Lot NumberJ04Y025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045616.06,045616.14
Lot NumberZ12F039 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045616.06,045616.14
Lot NumberL07W005 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045616.06,045616.14
Lot NumberP29H024 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045616.06,045616.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.