missing translation for 'onlineSavingsMsg'
Learn More
Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11389508

Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11389508
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11389508

Brand: Thermo Scientific Alfa Aesar 043340.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For vacuum deposition.Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]

Specifications

Boiling Point 707°C
Melting Point 300°C
Color Yellow
Physical Form Fused Pieces
Quantity 5 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 246.03
Odor Like rotten eggs
Density 3.43 g/cm3
Chemical Name or Material Arsenic(III) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN18L025 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number043340.14,043340.06
Lot NumberR29K024 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number043340.14,043340.06
Lot NumberR11I039 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number043340.14,043340.06
Lot NumberM26D030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043340.14,043340.06
Lot NumberX11H066 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043340.14,043340.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.