missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

For vacuum deposition

£117.59 - £459.29

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364977
View Documents
Thermo Scientific Alfa Aesar
041980.06
5 g
£117.59
5g
Please to purchase this item. Need a web account? Register with us today!
11354977
View Documents
Thermo Scientific Alfa Aesar
041980.14
25 g
£459.29
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-33-9
246.02
CUGMJFZCCDSABL-UHFFFAOYSA-N
[S--].[S--].[S--].[As+3].[As+3]
As2S3
MFCD00003435
diarsenic(3+) trisulfanediide

Specifications

707°C
1303-33-9
As2S3
UN1557
CUGMJFZCCDSABL-UHFFFAOYSA-N
diarsenic(3+) trisulfanediide
246.03
Like rotten eggs
(metals basis)
300°C
Fused Lump
MFCD00003435
14,806
[S--].[S--].[S--].[As+3].[As+3]
246.02
99.999%
3.43 g/mL
Arsenic(III) sulfide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-117-4

missing translation for 'rtecsNumber' : CG2638000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only