missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

£136.68 - £523.61

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442337
View Documents
Thermo Scientific Alfa Aesar
045522.06
5 g
£136.68
5g
Please to purchase this item. Need a web account? Register with us today!
15452337
View Documents
Thermo Scientific Alfa Aesar
045522.14
25 g
£523.61
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-33-9
246.02
CUGMJFZCCDSABL-UHFFFAOYSA-N
[S--].[S--].[S--].[As+3].[As+3]
As2S3
MFCD00003435
diarsenic(3+) trisulfanediide

Specifications

707°C
1303-33-9
99.99% (Metals basis)
MFCD00003435
CUGMJFZCCDSABL-UHFFFAOYSA-N
diarsenic(3+) trisulfanediide
246.03
Arsenic(III) sulfide
300°C
Powder
As2S3
Insoluble in water.
[S--].[S--].[S--].[As+3].[As+3]
246.02
3.43 g/cm3
Videos
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 215-117-4

missing translation for 'rtecsNumber' : CG2638000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only