missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) telluride, 99.999% (metals basis), Thermo Scientific Chemicals

£183.92 - £2262.00

Chemical Identifiers

CAS 12044-54-1
Molecular Formula As2Te3
Molecular Weight (g/mol) 532.643
MDL Number MFCD00014167
InChI Key GTIUFDICMGTSPM-UHFFFAOYSA-N
Synonym arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
PubChem CID 15770922
SMILES [As](=[Te])[Te][As]=[Te]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388848
View Documents
Thermo Scientific Alfa Aesar
041709.09
10 g
£183.92
10g
Please to purchase this item. Need a web account? Register with us today!
11378848
View Documents
Thermo Scientific Alfa Aesar
041709.18
50 g
£746.72
50g
Please to purchase this item. Need a web account? Register with us today!
15419189
View Documents
Thermo Scientific Alfa Aesar
041709.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12044-54-1
532.643
GTIUFDICMGTSPM-UHFFFAOYSA-N
15770922
As2Te3
MFCD00014167
arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
[As](=[Te])[Te][As]=[Te]

Specifications

381°C
Pieces
MFCD00014167
arsenic iii telluride, tellanylidenearsanyl tellanyl arsanetellone, arsenic tritelluride
[As](=[Te])[Te][As]=[Te]
15770922
99.999%
6.5
Arsenic(III) telluride
12044-54-1
As2Te3
UN1557
GTIUFDICMGTSPM-UHFFFAOYSA-N
532.643
532.64
Plastic bottle
6.5
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 234-955-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only