missing translation for 'onlineSavingsMsg'
Learn More
Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™ Product Code.: 11366698

Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™

Product Code. 11366698
100 g
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
1 kg
This item is not returnable. View return policy

Product Code. 11366698

Brand: Thermo Scientific Alfa Aesar 013375.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Color Gray
Physical Form Polycrystalline Lump
Quantity 100 g
Assay Percent Range 99.9999% (Metals basis)
Odor Odorless
Appearance Gray lump
Chemical Name or Material Arsenic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY30I045 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.