Learn More
Arsenic Standard, 1000μg/g in Base Oil 75, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ORG-AS8-2Z
Code : 04
Additional Details : Weight : 0.09000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 1000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Arsenic Oil Standard | |
Triple-leached LDPE bottle | |
99.9 | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00085309 | |
[As] | |
74.92 | |
CHEBI:27563 | |
Matrix Concentration Cat. No. Base Oil 20 1000μg/g ORG-AS8-2Z Base Oil 75 5000μg/g ORG-AS8-4Z |
7440-38-2,8042-47-5 | |
50 g | |
99.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
As | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
arsenic | |
5359596 | |
Oil/Plasma | |
Base Oil 75 |
Safety and Handling
- 1000 ug/g Arsenic in Base Oil 75
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : arsenic
Product intended for laboratory use only