Learn More
Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack
Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.
£144.00 - £202.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Product Code | Brand | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Tip Style | Fit for Purpose | Price | Quantity & Availability | ||
10049000
|
Thermo Scientific™
2065E |
ART 100E, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Sterile & PCR Ready |
£174.00
Pack of 960 |
In Stock
Log in to see stock. |
||
10577581
|
Thermo Scientific™
2070 |
ART 300, Filtered, Sterile, Lift-off Lid Rack | 300 μL | Micro Point | Sterile & PCR Ready |
£202.00
Pack of 960 |
|
||
10029040
|
Thermo Scientific™
2069 |
ART 200, Filtered, Sterile, Lift-off Lid Rack | 200 μL | Universal | Sterile & PCR Ready |
£176.00
Pack of 960 |
In Stock
Log in to see stock. |
||
10292761
|
Thermo Scientific™
2065 |
ART 100, Filtered, Sterile, Lift-off Lid Rack | 100 μL | Micro Point | Sterile & PCR Ready |
£175.00
Pack of 960 |
In Stock
Log in to see stock. |
||
12654870
|
Thermo Scientific™
2079E |
ART 1000E, Filtered, Sterile, Lift-off Lid Rack | 1000 μL | Universal | Sterile & PCR Ready |
£166.00
Pack of 800 |
In Stock
Log in to see stock. |
||
10098960
|
Thermo Scientific™
2139F |
ART 10F, Filtered, Sterile, Lift-off Lid Rack | 0.5 to 10 μL | Micro Point | Sterile & PCR Ready |
£175.00
Pack of 960 |
In Stock
Log in to see stock. |
||
10282761
|
Thermo Scientific™
2149P |
ART 20, Filtered, Sterile, Lift-off Lid Rack | 20 μL | Micro Point | Sterile & PCR Ready |
£177.00
Pack of 960 |
In Stock
Log in to see stock. |
||
10707811
|
Thermo Scientific™
2149E |
ART 10, Filtered, Sterile, Lift-off Lid Rack | 10 μL | Micro Point | Sterile & PCR Ready |
£144.00
Pack of 960 |
In Stock
Log in to see stock. |
||
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier pipette tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Filtered | |
Not Autoclavable | |
Lift-off Lid Rack | |
ART |
Sterile | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Universal Fit | |
Polypropylene |