Learn More
Thermo Scientific™ ART™ Barrier Specialty Pipette Tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Brand: Thermo Scientific™ 2350
62.47 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are certified free from RNase, DNase and pyrogen
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettetes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettetes and Thermo Scientific™ Finnpipettete™ Focus pipettetes

Specifications
20 μL | |
ART 20 ERGO-F | |
5.08cm | |
Rack | |
Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus | |
Sterile and PCR Ready |
Sterile | |
2 in. | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Ergonomic | |
ART |
Special Offers
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.