missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Gel Loading Pipette Tips
GREENER_CHOICE

Product Code. 10028990
Click to view available options
Volume (Metric):
20 μL
100 μL
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Pack of 960

Product Code. 10028990

Brand: Thermo Scientific™ 2155P

Only null left
Add to basket
Add to basket

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Gel Loading Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Gel loading for loading acrylamide or agarose gels

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY

Specifications

Description ART Gel 20P, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Model ART Gel 20P
Length (English) 2.79 in.
Length (Metric) 7.09cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Gel Loading
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.