missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette tips
GREENER_CHOICE

Product Code. 10008970
Click to view available options
Volume (Metric):
0.5 to 20 μL
10 μL
1000 μL
200 μL
50 μL
Model:
ART 10 REACH
ART 1000
ART 200U
ART 20E
ART 50U
Tip Style:
Ergonomic
Extended Length
Micro Point
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Pack of 800
Pack of 960

Product Code. 10008970

Brand: Thermo Scientific™ 2149

Only null left
Add to basket
Add to basket

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips DNase, RNase, DNA, ATP, PCR Inhibitor, Endotoxin, and Pyrogen Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to Aid in Ppipetteting for a Variety of Applications:

Extended length: to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading: for loading acrylamide or agarose gels

Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials

Solvent safe: for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY

Specifications

Description ART 20E, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 0.5 to 20 μL
Filtered Filtered
Sterility Sterile
Model ART 20E
Length (English) 1.78 in.
Length (Metric) 4.521cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Micro Point
For Use With (Equipment) Eppendorf
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.