Learn More
Thermo Scientific™ ART™ Wide Bore Filtered Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Brand: Thermo Scientific™ 2079G
112.77 GBP valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
- Sterile tips are certified free from RNase, DNase and pyrogen
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
ART 1000G, Filtered, Sterile, Lift-off Lid Rack | |
Filtered | |
ART 1000G | |
8.54cm | |
Lift-off Lid Rack | |
Universal Fit | |
Sterile & PCR Ready |
1000 μL | |
Sterile | |
3.36 in. | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Genomic/Wide Orifice | |
ART |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.