missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Articulating Arm Boom Stand on Base

Articulating arms provide great freedom of movement and positioning. Motic™ Articulating Arm Boom Stand on Base permits large, fine, stable and fluid microscope positioning.

Brand:  Motic™ 1101010100074

Product Code. 17118648

  • £487.00 / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Large, fine, stable, and fluid microscope positioning
  • Ideal option for the establishment of a spot and/or quick inspection station
  • Articulating arm movement combined with a fast release locking mechanism permits multiple observation points for a sample or the use of one stand for samples of various sizes
  • Swiveling capacity is convenient for removing samples from the work area
  • Suitable for a variety of microscopes
  • Head holder required
Specifications

Specifications

Articulating Arm Boom Stand Microscope
Pole diameter: 32 mm, Mounting pole diameter: 32 mm
445 mm Base, 400 mm Pole
Head holder required
Optical center (max): 780 mm
SFC-11 / SMZ-140 / SMZ-161 / SMZ-168 / SMZ-171 / K Series Microscopes
300 mm Base
300 mm Base
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.