missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH AS 200 Standard Clamping Devices
£645.00 - £845.00
Specifications
Product Type | Clamping Devices |
---|---|
Diameter (Metric) | 100/150/200/203 mm |
For Use With (Equipment) | Analytical sieve shaker AS 200 |
Packaging Quantity | 1 Ea. |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
10136791
|
RETSCH
32.662.0005 |
Dry Sieving |
£645.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
10789902
|
RETSCH
32.662.0007 |
Wet Sieving |
£845.00
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Specifications
Clamping Devices | |
100/150/200/203 mm | |
Analytical sieve shaker AS 200 | |
1 Ea. |