Learn More
Thermo Scientific™ ASAPath™ COVID-19 Antigen Rapid Test (CE-IVD)
Obtain COVID-19 test results fast with the easy-to-use, disposable Thermo Scientific ASAPath COVID-19 Antigen Rapid Test.
Brand:  Thermo Scientific™ 10028730
Additional Details : Weight : 0.20000kg
Product Code. 17286053
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Obtain COVID-19 test results within 30 minutes with the fast, easy-to-use, disposable Thermo Scientific™ ASAPath™ COVID-19 Antigen Rapid Test. This test is a qualitative lateral flow immunoassay intended to detect the nucleocapsid protein from the SARS-CoV-2 virus, which causes COVID-19, in nasal swab samples from symptomatic individuals who are suspected of COVID-19 by a healthcare professional, or individuals with or without symptoms or other epidemiological reasons to suspect COVID-19 infection.
  • Rapid Antigen Test for COVID-19
  • Detect nasal swab sample
  • Kit contains 10 sets of tests
  • Easy nasal swab sample collection
  • Test can be done by a single person
  • Results within 30 minutes
  • Easy-to-read results
  • No special reader necessary
  • Antibody pool targeting several virus protein regions for variant detection
  • Single use, disposable device
Specifications
Specifications
Room Temperature
ASAPath™ COVID-19 Antigen Rapid Test (CE-IVD)
One kit contains ten (10) Antigen Rapid Tests, each of which includes cassette, swab, extraction tube, dropper cap, and tube stand
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • X10 ASAPath Covid-19 Antigen Rapid Test
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hexadecan-1-yl(trimethyl)ammonium chloride
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product is CE Marked but not 510(k) cleared nor authorized for Emergency Use in US and not yet available for sale in the U.S. Availability of the product in each country depends on local regulatory marketing authorization status.