missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Fine Granular, 20 to 80 Mesh, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
125 g
500 g
1 kg
Packaging:
Poly Bottle
Specifications
Specifications
Mesh Size | 20 to 80 Mesh |
Packaging | Poly Bottle |
Quantity | 500 g |
Specific Gravity | 1L = 1.65kg |
Formula Weight | 176.12 |
Grade | FCC/USP |
Physical Form | Fine Granular |
Chemical Name or Material | Ascorbic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction