missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ascorbic Acid, Fine Powder, 200 to 325 Mesh, Multi-Compendial, U.S.P., J.T. Baker™
£876.00 - £1760.00
Chemical Identifiers
CAS | 50-81-7 |
---|---|
Molecular Formula | C6H8O6 |
Molecular Weight (g/mol) | 176.12 |
MDL Number | MFCD00064328 |
InChI Key | CIWBSHSKHKDKBQ-DOMZIZNONA-N |
Synonym | l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin |
PubChem CID | 54670067 |
ChEBI | CHEBI:29073 |
IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
SMILES | OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15527884
|
Avantor J.T.Baker
0939-07 |
500 g | Wide Mouth Amber Glass Bottle |
£876.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15537884
|
Avantor J.T.Baker
0939-08 |
1 kg | Wide Mouth Amber Glass Bottle |
£1760.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
50-81-7 | |
176.12 | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
54670067 | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
C6H8O6 | |
MFCD00064328 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
CHEBI:29073 | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O |
Specifications
50-81-7 | |
C6H8O6 | |
1L = 1.65kg | |
CIWBSHSKHKDKBQ-DOMZIZNONA-N | |
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one | |
54670067 | |
176.12 | |
Fine Powder |
200 to 325 Mesh | |
MFCD00064328 | |
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin | |
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O | |
176.12 | |
CHEBI:29073 | |
Multi-Compendial/USP | |
Ascorbic Acid |