missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Ashing Furnace L 40/11 BO

Specially designed for processes that require the incineration of large sample quantities. Ideal for applications such as ashing of food, thermal cleaning of injection molding tools, or the determination of annealing loss.

Brand:  Nabertherm™ LB401K1ON1

Product Code. 17367919

  • £12990.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spring-assisted closing of the furnace door (flap door) with mechanical locking prevents unintentional opening
  • Ceramic heating plates with embedded heating wire
  • Steel collecting pan protects the bottom insulation
  • Inlet-air is preheated through the bottom heating plate
  • Exhaust gas fan extracts flue gases from the furnace and simultaneously supplies fresh air to the furnace so that sufficient oxygen is always available for the incineration process
  • Over-temperature limiter with adjustable cutout temperature to protect the oven and load
  • Controller C550 (10 programs with each 20 segments), alternative controllers are available
  • Tmax: 600°C (Incineration Process), 1100°C (Subsequent Process)
  • Temperature control of post combustion can be set up to 850°C
Specifications

Specifications

C550
600°C (Incineration Process), 1100°C (Subsequent Process)
Flap Door
825 x 530 x 800 mm
3 Phase
11.5 kW
40 L
Furnace
490 x 320 x 250 mm
110 kg
400 V
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.