missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aspartame, 98%

Product Code. p-7042156
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15444109

Brand: Thermo Scientific Chemicals J61523.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

A non-nutritive sweetener and dipeptide ester about 160 times sweeter than sucrose | CAS: 22839-47-0 | C14H18N2O5 | 294.31 g/mol

Aspartame is a synthetic and low-calorie sweetener and is used as an active ingredient in consumer foods, beverages, soft drinks, breath mints, sugar free chewing gum, cocoa mixes, frozen desserts, chewable vitamin supplements, milk drinks, cerals, pharmaceutical drugs and supplements, tabletop sweeteners and diet sodas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aspartame is a synthetic and low-calorie sweetener and is used as an active ingredient in consumer foods, beverages, soft drinks, breath mints, sugar free chewing gum, cocoa mixes, frozen desserts, chewable vitamin supplements, milk drinks, cerals, pharmaceutical drugs and supplements, tabletop sweeteners and diet sodas.

Solubility
Soluble in formic acid, dimethyl sulfoxide. Sparingly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Melting Point 243°C to 245°C
Color White
Linear Formula HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
Beilstein 2223850
Merck Index 14,839
Solubility Information Soluble in formic acid,dimethyl sulfoxide. Sparingly soluble in water and ethanol.
Quantity 5 g
Formula Weight 294.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Aspartame
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.