Learn More
Grant Instruments™ Replacement Hydrophobic Filter
Prevents contaminants from entering the pump.
Brand: Grant Instruments™ FA-1
Description
- For use with Grant FTA-1 Aspirator
- 0.027 micron in-line filter prevents contaminants from entering the pump
- Direction of flow indicated on filter body for correct installation
Specifications
FTA-1 Aspirator |
Product Content Correction
Product Title
Grant Instruments™ Replacement Hydrophobic Filter > Replacement hydrophobic filter
Spot an opportunity for improvement?Share a Content Correction