missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Assay Kit

Brand:  Macherey-Nagel™ 935019


Product Code. 10014530

  • £22.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Colorimetric Analysis or Titrimetry
1 Each
0.25 to 2.0 mg/L Cl2
Assay Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • VISOCOLOUR alpha chlorine

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : N,N-diethyl-1,4-phenylene diammonium sulphate, sulphuric acid

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Assay Kit > Range: 0.25 to 2.0mg/L Cl2

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.