missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Square Cover Glasses for Microscopy, Selected Thickness

Preserve and protect specimens on slides. Karl Hecht™ Assistent™ Square Cover Glasses for Microscopy, Selected Thickness are manufactured from a super-transparent borosilicate glass, preventing virtually any color distortion.

Brand:  Karl Hecht™ 41014521

 View more versions of this product

Product Code. 10573462

  • £26.25 / Pack of 100
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 0.17 ± 0.01mm thickness
  • Excellent flatness
  • With CE marking
  • Optimum resistance to chemical attack
  • Refractive index finely adapted to microscopes
  • High spectral transmission
  • Excellent internal glass quality with very low levels of inclusions, striae, bubbles, streaks, etc.
Specifications

Specifications

Coverslip
0.17 ±0.01 mm
CE
Glass
24 x 24 mm
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Hecht™ Assistent™ Square Cover Glasses for Microscopy, Selected Thickness > Size: 24 x 24mm; Thickness: 0.17 ± 0.01mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.