missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Assorted Cork Stoppers

A simple and economical option for closing tubes, flasks and other narrow-necked vessels in the laboratory.

Brand:  Fisherbrand™

Code : 02

Additional Details : Weight : 0.27000kg

Product Code. 12346477

  • £75.70 / Pack of 144
Estimated Shipment: 15-11-2024
to see stock.
Add to basket

Description

Description

  • 24 each of: 9mm, 12mm, 16mm, 21mm and 22mm bottom diameters
  • 6 each of: 25.5mm, 28.5mm, 32mm and 35mm
Specifications

Specifications

Cork Stopper
Cork
Tapered
144/Pk.
Narrow-necked vessels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers