Learn More
ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™
ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
Brand: Paragon Scientific 134290
Additional Details : CAS Number : 8042-47-5 Weight : 0.90000kg
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Specifications
500 mL | |
Color Reference Standard | |
Certified according to ISO 17025 / ISO 17034 under UKAS accreditation |
Safety and Handling
- ASTM Colour Reference Standard
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H350-May cause cancer.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : petroleum distilates
Nominal values quoted