missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE ASTM Specific Gravity Hydrometer
The ASTM Specific Gravity hydrometer is manufactured to the specifications of the American Society for Testing and Materials.
Brand: VEE GEE 67130H
This item is not returnable.
View return policy
Description
- Approx. Length: 330 mm
- Calibrated for use at 15.5°C/60°F
Specifications
Specific Gravity Hydrometer | |
0.001 | |
ASTM Specific Gravity Hydrometer, ASTM 130H, 1.250 to 1.300 | |
1.250 to 1.300 |
130H | |
26 cm | |
Glass | |
15.5°C (60°F) |