missing translation for 'onlineSavingsMsg'
Learn More

AT 101, Tocris Bioscience™

Product Code. 18095812
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Downregulates Bcl-2 and Mcl-1; pro-apoptotic

R-(-)-enantiomer of gossypol (Cat. No. 1964). Mimics the BH3 domains of Bcl-2, Bcl-XL and Mcl-1. Disrupts heterodimerization of Bcl-2 with proapoptotic family members. Induces apoptosis in vitro through activation of caspase-9; cytotoxic to multiple myeloma and drug-resistant cell lines. Delays onset of androgen-independent growth of VCaP prostate cancer xenografts in vivo.

Chemical Identifiers

CAS 90141-22-3
Molecular Formula C30H30O8
Molecular Weight (g/mol) 518.562
InChI Key QBKSWRVVCFFDOT-UHFFFAOYSA-N
Synonym gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
PubChem CID 3503
ChEBI CHEBI:28584
IUPAC Name 7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C

Specifications

Quantity 50 mg
Formula Weight 518.56
Percent Purity >98%
Chemical Name or Material AT 101
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.