missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ ATAS Optic Split / Splitless FocusLiner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092272
This item is not returnable.
View return policy
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with ATAS Optic instruments
Specifications
5/Pk. | |
Split/Splitless Focusliner Inlet Liner | |
5 mm |
81 mm | |
3 mm |