missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ ATAS Optic Split / Splitless FocusLiner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09227201

 View more versions of this product

Product Code. 12924091

  • £29.95 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with ATAS Optic instruments
Specifications

Specifications

1 Ea.
Split/Splitless Focusliner Inlet Liner
5 mm
81 mm
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers