missing translation for 'onlineSavingsMsg'
Learn More

ATF6 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for ATF6

Brand:  Abnova H00022926-T01.100uL

Product Code. 16166752

  • £182.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ATF6 is an endoplasmic reticulum (ER) stress-regulated transmembrane transcription factor that activates the transcription of ER molecules.

  • Activating transcription factor 6
  • Transfected cell line: 293T
  • Plasmid: pCMV-ATF full-length
  • Host: Human
  • Theoretical molecular weight: 22.33kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

Specifications

293T
ATF6 293T Cell Transient Overexpression Lysate(Denatured)
22.33
pCMV-ATF6 full-length
100 μL
Human
Western Blot
BC014969.1
6.8
Transient overexpression cell Lysate was tested with Anti-ATF6 antibody (H00022926-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers