Learn More
ATG16L1 Mouse anti-Human, Unconjugated, Clone: 1G5G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67943-1-IG-20UL
This item is not returnable.
View return policy
Description
The ATG16L1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATG16L1. This antibody recognizes human, mouse, rat antigen. The ATG16L1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ATG16L1 | |
Monoclonal | |
Unconjugated | |
20 μL | |
55054 | |
IgG1 |
Western Blot, ELISA | |
1G5G7 | |
Mouse | |
Primary | |
Human, Rat, Mouse |