missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Athletic All-Round Assembly Gloves

Product Code. 17210341
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, dirt-resistant gloves are constructed from a blend of polyamide and elastane, with an NBR foam coating for enhanced grip capabilities.

  • Slim-fit provides tactile feedback when assembling smaller workpieces
  • NBR foam coating is abrasion resistant and enhances grip in dry or slightly damp environments
  • Dirt-resistant shell is ideal for mechanical tasks
  • Certified by OEKO-TEX™ Standard 100
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material NBR Foam
Color Gray, Black
Material Elastane, Polyamide
Size 6
EN388 Standard 4122X
For Use With (Application) Assembly, Packaging, Precision Work, and Transportation
Gender Unisex
Glove Cuff Style Knit
Series Athletic All-Round
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'