Learn More
Uvex™ Athletic All-Round Safety Glove
Click to view available options
Size: 11
Description
- Outer material made from polyamide and elastane
- Robust NBR foam coating
- Good mechanical abrasion resistance thanks to the robust NBR foam coating
- Good grip in dry and slightly damp areas
- Perfect fit thanks to the 'slim fit' design and elastane in the liner
- Safety glove with knitted cuff
Specifications
Certifications/Compliance | EN 388 (4 1 2 2 X) |
Coating Material | Nitrile Foam |
Color | Gray, Black |
Material | Elastane, Polyamide |
Size | 11 |