Learn More
Uvex™ Athletic Lite ESD Safety Glove
Click to view available options
Size:
6
7
8
9
10
11
Description
- Light and breathable safety glove
- Particularly flexible and sensitive glove for multiple areas of application
- Touchscreen capability
- Certified in line with Oeko-Tex Standard 100
- Good grip in dry and slightly damp areas
- High breathability thanks to open-pored coating that reduces perspiration inside the glove
- Good tactile feel when carrying out precision work
- Perfect fit thanks to the 'slim fit' design and elastane in the liner
Specifications
Certifications/Compliance | DIN EN 16350, EN 388 (4121X) |
Coating Material | NBR MicroFoam |
Color | Blue, Anthracite |
Material | Polyamide, Carbon Fiber, Elastane |
Size | 6 |