Learn More
Biochrom Atlantis 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines
The Biochrom Asys Atlantis microplate washer for standard ANSI SBS footprint 96-well plates with flexiblility in washing options and simple operation. It is the perfect choice to compliment any microplate reader in any applications environment.
Brand:  Biochrom G021101
Product Code. 15531913
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Configurable to many different well formats for maximum flexibility
  • Optional 16-way manifold for washing 384-well plates
  • Detachable plate carrier for easy cleaning
  • Low noise, pressure-free aspiration and dispensing pumps
  • Auto rinse function prevents liquid path clogging
  • Reduces aerosols due to cover and pumping technology
  • Optional liquid level sensors
Specifications
Specifications
Atlantis 2 Microplate Washer for 96 Well Plates with 2 Liquid Lines
96-well Plate, 384-well Plate
6 kg
ELISA
46 x 21 x 21 cm
100/240 V, 180/250 V autosensing
50 to 2000 μL
±1 μL/well
2 x 16 character display with backlight
50 μL
14 lb.