missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Atlas™ Evacuable Pellet Die 40mm
Brand: Specac™
Code : Z3
Additional Details : Weight : 5.70000kg
Description
40mm Evacuable Pellet Die used for compacting powdered samples into discs or briquettes with use of a pressing system. Well suited for KBr pellet production for FTIR analysis and XRF pellets. This product comprises of a base, body, plunger, pair of pressing surfaces, extractor ring + O-Ring Kit.Specifications
40 mm |