missing translation for 'onlineSavingsMsg'
Learn More
Bollé Safety Atom Platinum™ Safety Goggles Product Code.: 12679090
In Stock
to see stock.
Add to basket

Bollé Safety Atom Platinum™ Safety Goggles

Product Code. 12679090
ATOEPSI (Sealed)
Click to view available options
Model:
ATOAPSI (Ventilated)
ATOEDEPSI (Sealed Double Lens)
ATOEPSI (Sealed)
ATOFAPSI (Breathable Foam)
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 12679090

Brand: Bollé Safety ATOEPSI

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Compact, lightweight and extremely comfortable, ATOM model safety goggles offer enhanced protection

  • Frame material: blue thermoplastic rubber
  • Lens material: clear polycarbonate
  • Lense coating: Platinum™ anti-fog / anti-scratch coating
TRUSTED_SUSTAINABILITY

Specifications

Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety goggles
Frame Color Blue
Includes Sealed
Model ATOEPSI (Sealed)
No. per Pack 1 Ea.
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Thermoplastic Rubber
UV Protection Yes
Show More Show Less
  • Bolle Safety Visor ATOM
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Bolle Safety Visor ATOM

    • Bollé Safety - Platinum coating
      Video Player is loading.
      Current Time 0:00
      Duration 1:06
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:06
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected

      Bolle Safety Platinum coating

    • Bollé Safety - Platinum efficiency demonstration
      Video Player is loading.
      Current Time 0:00
      Duration 3:09
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 3:09
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected

      Bolle Safety Platinum efficiency demonstration

    Specification Sheets

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Bollé Safety Atom Platinum™ Safety Goggles >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.