missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atomoxetine hydrochloride, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 82248-59-7 |
Packaging | Glass Bottle |
Quantity | 1 g |
Color | White to Beige |
Certificate of Analysis | Available |
Molecular Formula | C17H22ClNO |
MDL Number | 06410992 |
InChI Key | LUCXVPAZUDVVBT-UNTBIKODSA-N |
SMILES | [H+].[Cl-].CNCC[C@@H](OC1=CC=CC=C1C)C1=CC=CC=C1 |
IUPAC Name | hydrogen methyl[(3R)-3-(2-methylphenoxy)-3-phenylpropyl]amine chloride |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'