Learn More
ATP Solution (10 mM)
ATP Solution (10 mM)
Brand: Invitrogen PV3227
Code : 50
Additional Details : Weight : 0.50000kg
Description
ATP is an essential component for kinase assays using LanthaScreen™ Technology. Invitrogen's ATP is 10mM in water.Specifications
ATP is shipped on dry ice and should be stored at -20°C. | |
Dry Ice | |
Unlabeled |
Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
10 mM | |
500 μL |
For Research Use Only. Not for use in diagnostic procedures.