Learn More
ATP1B1 Monoclonal Antibody (M17-P5-F11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA3930
Description
MA3-930 detects sodium/potassium ATPase beta 1 from mouse, human, sheep, canine, and pig tissues. This antibody does not recognize the beta 1 or beta 2 subunit in rat. MA3-930 has been successfully used in Western blot and immunohistochemistry and immunocytochemistry procedures. By Western blot, this antibody detects an ∽42 kDa protein in liver samples and ∽55 kDa in kidney samples representing beta sodium/potassium ATPase in sheep liver extract. Immunohistochemical staining of the sodium/potassium ATPase with MA3-930 yields a pattern consistent with plasma membrane localization. The MA3-930 antigen is purified lamb kidney sodium/potassium ATPase beta. This antibody recognizes and epitope between amino acid residues 195-199 of sheep sodium/potassium ATPase beta 1.
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 1 subunit. Alternatively spliced transcript variants encoding different isoforms have been described, but their biological validity is not known.Specifications
ATP1B1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P05026, P05027, P05028, P06583, P14094 | |
ATP1B1 | |
Purified lamb kidney beta sodium/potassium ATPase. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Ovine, Porcine | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
M17-P5-F11 | |
Unconjugated | |
ATP1B1 | |
adenosinetriphosphatase; ATP1B; ATP1B1; Atp4b; ATPase beta subunit; ATPase Na+/K+ transporting subunit beta 1; ATPase, Na+/K+ transporting, beta 1 polypeptide; Atpb; Atpb-1; ATPBS; Beta 1-subunit of Na(+),K(+)-ATPase; hypothetical protein LOC519758; MGC1798; Na, K-ATPase beta-1 polypeptide; Na,K-ATPase beta 1 subunit; Na+, K+-ATPase; Na+, K+-ATPase beta-subunit; Na+,K+-ATPase beta 1 subunit; Na+/K+ ATPase Beta; NKbeta1; OTTHUMP00000032537; OTTHUMP00000032538; P-type ATPase; Sodium Potassium ATPase; sodium pump subunit beta-1; sodium/potassium ATPase beta subunit; sodium/potassium-dependent ATPase beta-1 subunit; sodium/potassium-dependent ATPase subunit beta-1; sodium/potassium-transporting ATPase beta-1 chain; sodium/potassium-transporting ATPase subunit beta-1; sodium-potassium ATPase subunit beta 1 (non-catalytic); unnamed protein product | |
Mouse | |
Protein A | |
RUO | |
11931, 396898, 403966, 443384, 481 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |