missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5D Mouse anti-Human, Clone: 1G12C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66673-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.Specifications
ATP5D | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30049 | |
ATP5D | |
ATP5D Fusion Protein Ag7016 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1G12C7 | |
Unconjugated | |
ATP5D | |
ATP5D, F ATPase delta subunit | |
Mouse | |
Protein G | |
RUO | |
513 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction