missing translation for 'onlineSavingsMsg'
Learn More

ATR Monoclonal Antibody (3G6D2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA548382

Product Code. 17925821

  • £396.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATR
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q13535
ATR
Purified recombinant fragment of human ATR (AA: 366-550) expressed in E. Coli.
100 μg
Primary
Human
Liquid
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
3G6D2
Unconjugated
ATR
2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8
Mouse
Protein G
RUO
545
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers