Learn More
3M™ Attest™ Rapid Readout Biological Indicators
Monitors the effectiveness of the steam sterilization process and provides results within 3 hours. 3M™ Attest™ Rapid Readout Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
Brand:  3M™ 7000053661
Product Code. 12318199
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Easy to use and interpret with visual color readout
  • Self-contained to significantly reduce the possibility of contamination
  • Minimizes false positives
Specifications
Specifications
Autoclavable
Brown
50 Pk.
ISO 11138
Biological Indicator
Steam