Learn More
Applied Biosystems™ Attune™ Flow Cytometer Syringe
The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
Brand:  Applied Biosystems™ 4452079
375.69 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
 View more versions of this product
Product Code. 15830413
  • £410.00 / 1mL
Explore more special offers
Description
Description
  • The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attune™
1 mL
Attune Acoustic Focusing Cytometer
Store at room temperature
Room Temperature
Syringe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ Attune™ Flow Cytometer Syringe > 1 mL sample syringe