Learn More
Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid (1X)
Brand: Invitrogen 4488621
28.32 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.
Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.
High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.

Specifications
Store at room temperature. | |
Liquid | |
1 L |
Attune™ Acoustic Focusing Cytometer | |
Attune™ | |
Focusing Fluid |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Attune Focusing Fluid (1X)
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl- 3(2H)-isothiazolone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6177142 | Certificate of Analysis | 10/02/2025 | 4488621 |
6112949 | Certificate of Analysis | 16/07/2024 | 4488621 |
3523462 | Certificate of Analysis | 01/09/2023 | 4488621 |
3638340 | Certificate of Analysis | 13/07/2023 | 4488621 |
3646375 | Certificate of Analysis | 31/05/2023 | 4488621 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.