missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ Focusing Fluid (1X)
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
Unit Size:
1 L
10 L
20 L
Pack of 6
Description
- An optimized buffer solution used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system
- This sterile, buffered, azide-free support reagent contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer
- Designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging
- Ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
- Convenient 20 L packaging provides enough focusing fluid to operate for at least 20 days (the average focusing fluid consumption per day is less than 1 L)
- Spigot helps to simplify the filling of onboard fluidics tanks
- Each batch of Attune™ Focusing Fluid is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer
Specifications
Specifications
| For Use With (Equipment) | Attune™ Acoustic Focusing Cytometer |
| Format | Liquid |
| Product Line | Attune |
| Quantity | 20 L |
| Product Type | Focusing Fluid |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction